Learn More
IFN gamma Monoclonal Antibody (MD-1), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$414.00 - $3285.00
Specifications
Antigen | IFN gamma |
---|---|
Clone | MD-1 |
Concentration | 1 mg/mL |
Applications | ELISA, Functional Assay |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
16-731-785
![]() |
Invitrogen™
16731785 |
500 μg |
Each for $414.00
|
|
|||||
16-731-738
![]() |
Invitrogen™
16731738 |
5 mg |
Each for $3,285.00
|
|
|||||
Description
The MD-1 antibody reacts with human, rhesus, cynomolgus and baboon interferon-gamma (IFN gamma), the signature Th1 cytokine, which is produced mainly by activated T cells, NK cells and B cells. IFN gamma is a dimeric protein. IFN gamma has antiviral and antiparasitic activities and also inhibits the proliferation of normal and transformed cells.
IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.Specifications
IFN gamma | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.2 | |
3458 | |
Primary | |
4°C | |
IFNG |
MD-1 | |
ELISA, Functional Assay | |
Functional Grade | |
Mouse | |
Human, Monkey | |
P01579 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.