Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-gamma Mouse anti-Hamster, Clone: MTH21, Novus Biologicals™

Mouse Monoclonal Antibody
$286.00 - $1033.00
Specifications
Antigen | IFN-gamma |
---|---|
Clone | MTH21 |
Concentration | 0.5 mg/ml |
Applications | ELISA |
Classification | Monoclonal |
Description
IFN-gamma Monoclonal antibody specifically detects IFN-gamma in Hamster samples. It is validated for ELISA, ELISPOTSpecifications
IFN-gamma | |
0.5 mg/ml | |
Monoclonal | |
Purified | |
RUO | |
Hamster | |
IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
Recombinant hamster (Mesocricetus auratus golden hamster) IFN-gamma | |
Primary | |
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
MTH21 | |
ELISA | |
Unconjugated | |
Mouse | |
Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers | |
0.2 um filtered PBS | |
3458 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title