Learn More
IFN gamma Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
$250.00 - $319.00
Specifications
Antigen | IFN gamma |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
500P3250UG
![]() |
Invitrogen™
500P3250UG |
50 μg |
Each for $250.00
|
|
|||||
500P32100UG
![]() |
Invitrogen™
500P32100UG |
100 μg |
Each for $319.00
|
![]() |
|||||
Description
AA Sequence of recombinant protein: MQDPYVKEAE NLKKYFNAGH SDVADNGTLF LGILKNWKEE SDRKIMQSQI VSFYFKLFKN FKDDQSIQKS VETIKEDMNV KFFNSNKKKR DDFEKLTNYS VTDLNVQRKA IHELIQVMAE LSPAAKTGKR KRSQMLFQGR RASQ. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IFN-gamma. Anti-Human IFN-gamma-specific antibody was purified by affinity chromatography employing an immobilized Human IFN-gamma matrix. Sandwich ELISA: To detect Human IFN-gamma by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human IFN-gamma (500-P32Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human IFN-gamma. Western Blot: To detect Human IFN-gamma by Western Blot analysis this antibody can be used at a concentration of 0.1- 0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human IFN-gamma is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.Specifications
IFN gamma | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P01579 | |
3458 | |
E.coli-derived Recombinant Human IFN-gamma. | |
Antigen Affinity Chromatography | |
IFNG |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; IFG; IFI; IFN gamma; IFN ?; Ifng; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFN?; Immune interferon; Interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon ?; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferon?; M-IFN-g; R-IFN-g | |
IFNG | |
Primary | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.