Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-gamma R1/CD119 Antibody (RM0093-7K23) - Azide and BSA Free, Novus Biologicals™
SDP

Rat Monoclonal Antibody

$206.50 - $560.50

Specifications

Antigen IFN-gamma R1/CD119
Clone RM0093-7K23
Concentration LYOPH
Dilution Western Blot 1:500-1:1000, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018431
SDP
View Documents
Novus Biologicals
NBP1224080.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP122408
SDP
View Documents
Novus Biologicals
NBP122408
0.1 mg
Each for $560.50
Only null left
Add to Cart
 
Description

Description

IFN-gamma R1/CD119 Monoclonal specifically detects IFN-gamma R1/CD119 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

IFN-gamma R1/CD119
LYOPH
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rat
Mouse
3459
IgG2
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
RM0093-7K23
Western Blot 1:500-1:1000, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
AVP, type 2, CD119, CD119 antigen, FLJ45734, IFN-gamma receptor 1, IFN-gamma-R1, IFNGR, immune interferon receptor 1, interferon gamma receptor 1, interferon-gamma receptor alpha chain, type 2
IFNGR1
Primary
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of INFGR1.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.