Learn More
IFN-gamma R1/CD119 Antibody (RM0093-7K23), DyLight 550, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP122408R
Catalog No. NBP122408R
Only null left
Add to Cart
Description
Description
IFN-gamma R1/CD119 Monoclonal specifically detects IFN-gamma R1/CD119 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
IFN-gamma R1/CD119
Monoclonal
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
IFNGR1
Mouse recombinant IFN-gamma R1/CD119
0.1 mL
Primary
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of INFGR1.
Store at 4C in the dark.
IgG2
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
RM0093-7K23
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
AVP, type 2, CD119, CD119 antigen, FLJ45734, IFN-gamma receptor 1, IFN-gamma-R1, IFNGR, immune interferon receptor 1, interferon gamma receptor 1, interferon-gamma receptor alpha chain, type 2
Rat
Protein A or G purified
Cytokine Research, Immunology, Innate Immunity, Myeloid derived Suppressor Cell, Neuroscience
3459
Mouse
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.