Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-gamma R1/CD119 Rabbit anti-Human, Clone: 10, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00
Specifications
Antigen | IFN-gamma R1/CD119 |
---|---|
Clone | 10 |
Dilution | Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
IFN-gamma R1/CD119 Monoclonal specifically detects IFN-gamma R1/CD119 in Human samples. It is validated for Western Blot, ELISA.Specifications
IFN-gamma R1/CD119 | |
Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
AVP, type 2, CD119, CD119 antigen, FLJ45734, IFN-gamma receptor 1, IFN-gamma-R1, IFNGR, immune interferon receptor 1, interferon gamma receptor 1, interferon-gamma receptor alpha chain, type 2 | |
Recombinant Human IFN-gamma R1/CD119 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
10 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Cytokine Research, Immunology, Innate Immunity, Myeloid derived Suppressor Cell, Neuroscience | |
0.2 um filtered solution in PBS | |
3459 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title