Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-gamma R1/CD119 Rabbit anti-Human, Mouse, Rat, Clone: 2B9Q1, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | IFN-gamma R1/CD119 |
---|---|
Clone | 2B9Q1 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
IFN-gamma R1/CD119 Monoclonal antibody specifically detects IFN-gamma R1/CD119 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
IFN-gamma R1/CD119 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cytokine Research, Immunology, Innate Immunity, Myeloid derived Suppressor Cell, Neuroscience | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3459 | |
IgG | |
Affinity purified |
2B9Q1 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
AVP, type 2, CD119, CD119 antigen, FLJ45734, IFN-gamma receptor 1, IFN-gamma-R1, IFNGR, immune interferon receptor 1, interferon gamma receptor 1, interferon-gamma receptor alpha chain, type 2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 270-489 of human IFN-gamma R1/CD119 (NP_000407.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title