Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN Induced GTPase-like Protein Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2441690.05ML
Description
IFN Induced GTPase-like Protein Polyclonal specifically detects IFN Induced GTPase-like Protein in Mouse samples. It is validated for Western Blot, ELISA.Specifications
IFN Induced GTPase-like Protein | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.5 M Sodium Chloride, pH 7.2 with 0.01% sodium azide | |
GM4841 | |
IFN Induced GTPase-like Protein Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a N-terminal region of mouse IFN Induced GTPase Like Protein. | |
Primary | |
Mouse | |
IgG |
Western Blot, ELISA | |
86.5 mg/mL | |
Western Blot 1:1000, ELISA 1:2000-1:10000 | |
EG225594, Ifgga3, interferon-gamma induced, Interferon-gamma-inducible GTPase Ifgga3 protein | |
Rabbit | |
0.05 mL | |
225594 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction