Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN Induced GTPase-like Protein Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990143
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0143 100 μL
50-199-0144 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0143 Supplier Rockland Immunochemicals Supplier No. 100401N95
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. A BLAST analysis was used to suggest cross-reactivity with IFN Induced GTPase Like Protein from mouse based on a 100% homology with the immunizing sequence. Reactivity against homologues from other sources is not known.

IFN Induced GTPase Like Protein is a novel protein involed in intracellular defense. It is suggested that IFN-induced GTPases may be an indicator for Lupus or other immunogenic abnormalities. Anti-IFN Induced GTPase Like Protein Antibody is ideal for researchers interested in autoimmune disease and apoptosis research.

Specifications

Antigen IFN Induced GTPase-like Protein
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 86.5 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.5M NaCl and 0.01% sodium azide; pH 7.2
Gene Gm4841
Gene Symbols Gm4841
Host Species Rabbit
Immunogen IFN Induced GTPase Like Protein was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a N-terminal region of mouse IFN Induced GTPase Like Protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225594
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.