Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-γ Mouse anti-Human, Rhesus, Cynomolgus, Baboon, R718, Clone: 4S.B3, BD Horizon™

Catalog No. BDB567060 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB567060 100 Tests
BDB567245 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BDB567060 Supplier BD Biosciences Supplier No. 567060
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFN-γ Monoclonal antibody specifically detects IFN-γ in Human, Rhesus, Cynomolgus, Baboon samples. It is validated for Intracellular staining (Flow Cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-γ
Applications Flow Cytometry
Classification Monoclonal
Clone 4S.B3
Conjugate R718
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias IFNG; Interferon-gamma; IFG; IFI; Type II interferon
Host Species Mouse
Immunogen Human IFN-γ from supernatants of S. aureus-stimulated PBMC
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Rhesus Macaque, Cynomolgus, Baboon
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.