Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-γ, Mouse Anti-Human, RY610, B27, BD Horizon™

Catalog No. BDB571143 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
Conjugate:
RY610
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
BDB571143 25 Tests RY610
BDB571142 100 Tests RY610
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB571143 Supplier BD Biosciences Supplier No. 571143
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IFN-γ Monoclonal antibody specifically detects IFN-γ in Human (QC Testing), Rhesus,Cynomolgus,Baboon (Tested in Development) samples. It is validated for Flow Cytometry (Intracellular Staining)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-γ
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone B27
Conjugate RY610
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias IFNG; Interferon-gamma; ; Type II interferon; MAF
Host Species Mouse
Immunogen Human IFN-γRecombinant Protein
Purification Method The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3458
Target Species Human, Rhesus Monkey, Cynomolgus, Baboon
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.