Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-γ Mouse anti-Human, V450, Clone: B27, BD

Catalog No. BDB560371 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
120 Tests
30 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560371 120 Tests
BDB560372 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB560371 Supplier BD Biosciences Supplier No. 560371
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The B27 antibody reacts with human interferon-γ (IFN-γ). This is a neutralizing antibody. The use of B27 antibody for epitope mapping of human IFN-γ has been described. The B27 antibody has been reported not to bind to denatured IFN-γ. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.

Host Species: Mouse
Clone: B27
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human IFN-γ Recombinant Protein

Immunocytochemistry, Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-γ
Applications Flow Cytometry
Classification Monoclonal
Clone B27
Conjugate Violet Fluor 450 (V450)
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human IFN-γ Recombinant Protein
Purification Method Affinity Purified
Quantity 120 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.