Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-omega/IFNW1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299256MFV500
Description
IFN-omega/IFNW1 Polyclonal antibody specifically detects IFN-omega/IFNW1 in Human samples. It is validated for ELISASpecifications
IFN-omega/IFNW1 | |
Polyclonal | |
ELISA | |
IFN-omega 1, interferon omega-1, Interferon alpha-II-1, interferon omega-1, interferon, omega 1 | |
Produced in rabbits immunized with purified, recombinant Human IFN-omega/IFNW1 (Accession#: P05000; Met1-Ser195) | |
0.1 mL | |
Cell Cycle and Replication | |
3467 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction