Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFN omega Monoclonal Antibody (OMG-7), eBioscience™
GREENER_CHOICE

Catalog No. 5017685
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-176-85 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-176-85 Supplier Invitrogen™ Supplier No. BMS1022
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: In the human genome, the IFN-omega gene family consists of seven members located on chromosome 9. However, only one of these genes is functional giving rise to the IFN-omega protein, whereas the others are non-functional pseudogenes. The IFN-omega gene is not expressed in unstimulated cells. Viral infection results in expression of the gene giving rise to the N-glycosylated protein consisting of 172 or 174 amino acids and an apparent molecular mass of about 25kDa. A single carbohydrate group consists mainly of biantennary complex oligosaccharides with variable amounts of N-acetyl neuraminic acid. In quantitative terms, IFN-omega is a major component of human leukocyte IFN; with a contribution to its total antiviral activity estimated to be in the range of 10 -15 %. Applications Tested: ELISA.

IFN-omega is a type I interferon that can be induced by virus-infected leukocytes. Members of the type I interferon family, which includes IFN-alpha, IFN-beta, and IFN-omega, signal through the IFNAR-1/IFNAR-2 receptor complex, and exert antiviral and antiproliferative activities. IFN-omega exhibits about 75% sequence homology with IFN-alpha, and contains two conserved disulfide bonds that are necessary for full biological activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN omega
Applications ELISA
Classification Monoclonal
Clone OMG-7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.2-7.4
Gene IFNW1
Gene Accession No. P05000
Gene Alias IFN o; IFN ω; IFNo; IFNomega; IFN-omega 1, interferon omega-1; IFNW; IFNW1; IFNω; Interferon; Interferon alpha-II-1; Interferon o; Interferon OMEGA; interferon omega 1; interferon omega-1; Interferon ω; interferon, omega 1
Gene Symbols IFNW1
Host Species Mouse
Immunogen Rec. Human IFN omega.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3467
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.