Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFN-γ Rat anti-Mouse, RB705, Clone: XMG1.2, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 570645
Description

Specifications
IFN-γ | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
15978 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Flow Cytometry (Intracellular Staining) | |
XMG1.2 | |
RB705 | |
Ifg; Ifng; IFN-γ; IFN-g; IFN-gamma; Interferon gamma; Type II Interferon | |
Mouse IFN-γ Recombinant Protein | |
100 μg | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction