Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFNA10 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987674
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7674 Supplier Bioss Supplier No. BS7024R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.

Specifications

Antigen IFNA10
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene IFNA10
Gene Accession No. P01566
Gene Alias IFN alpha; IFNA10; IFN-alpha-10; IFN-alphaC; interferon alpha 10; Interferon alpha-10; Interferon alpha-6L; Interferon alpha-C; interferon, alpha 10; LeIF C
Gene Symbols IFNA10
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human IFNA10/Interferon alpha 10.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3446
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.