Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFNA13 Monoclonal Antibody (01)
GREENER_CHOICE

Catalog No. PIMA528977
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528977 Supplier Invitrogen™ Supplier No. MA528977
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Cynomolgus IFNA13.

Interferon alpha (IFN alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component results in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. Interferon alpha is produced primarily by plasmacytoid dendritic cells and is a potent component of the anti viral innate immune response; it modulates adaptive immunity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFNA13
Applications ELISA
Classification Monoclonal
Clone 01
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias IFNA1; IFNA13; IFN-alpha 1b; IFN-alpha-1/13; Interferon; interferon alpha 13; interferon alpha 1b; interferon alpha-1/13; Interferon alpha-D; interferon, alpha 13; interferon-alpha1; leIF D
Host Species Mouse
Immunogen Recombinant Cynomolgus IFNA13 protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Cynomolgus Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.