Learn More
Invitrogen™ IFNA13 Monoclonal Antibody (3F9)

Description
Peptide Sequence: FTTKDSSAAW DEDLLDKFCT ELYQQLNDLE ACVMQEERVG ETPLMNADSI LAVKKYFRRI TLYLTEKKYS PCAWEVVRAE IMRSLSLSTN LQERLRRKE.
Specifications
Specifications
| Antigen | IFNA13 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3F9 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | IFNA13 |
| Gene Accession No. | P01562 |
| Gene Alias | IFNA1; IFNA13; IFN-alpha 1b; IFN-alpha-1/13; Interferon; interferon alpha 13; interferon alpha 1b; interferon alpha-1/13; Interferon alpha-D; interferon, alpha 13; interferon-alpha1; leIF D |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.