Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFNA14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597859
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597859 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597859 Supplier Invitrogen™ Supplier No. PA597859
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IFNA14 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFNA14
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene IFNA14
Gene Accession No. P01570
Gene Alias IFNA14; IFN-alpha-14; IFN-alphaH; Interferon; interferon alpha 14; interferon alpha-14; Interferon alpha-H; interferon lambda-2-H; interferon, alpha 14; leIF H; LEIF2H; OTTMUSG00000007662
Gene Symbols IFNA14
Host Species Rabbit
Immunogen Recombinant Human Interferon alpha-14 protein (24-189AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3448, 404549
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.