Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFNA7/IFNA16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103423
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103423 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103423 Supplier Invitrogen™ Supplier No. PA5103423
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IFNA7/16.

IFNA7 (Interferon Alpha 7) and IFNA16 (Interferon Alpha 16) are predicted to enable cytokine activity and type I interferon receptor binding activity and to be involved in several processes, including B cell activation; lymphocyte activation involved in immune response; and positive regulation of peptidyl-serine phosphorylation of STAT protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFNA7/IFNA16
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IFNA7
Gene Accession No. P01567, P05015
Gene Alias Interferon
Gene Symbols IFNA16, IFNA7
Host Species Rabbit
Immunogen A synthesized peptide derived from human IFNA16(Accession P05015), corresponding to amino acid residues G33-M83.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3444, 3449
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.