Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFNA16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545146
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545146 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545146 Supplier Invitrogen™ Supplier No. PA545146
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ILAVRKYFQR ITLYLMGKKY SPCAWEVVRA EIMRSFSFST NLQKGLRRKD Sequence homology: Human: 100%.

IFNA16 is produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFNA16
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene IFNA16
Gene Accession No. P05015
Gene Alias IFNA16; IFN-alpha-16; IFN-alpha-N-protein; IFN-alphaO; Interferon; interferon alpha 16; interferon alpha-16; Interferon alpha-WA; interferon, alpha 16
Gene Symbols IFNA16
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human IFNA16 (aa 140-189).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3449
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.