Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFNA17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103424
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103424 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103424 Supplier Invitrogen™ Supplier No. PA5103424
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IFN17.

Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFNA17
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IFNA17
Gene Accession No. P01571
Gene Alias IFNA; IFNA17; IFN-alpha-17; IFN-alphaI; INFA; Interferon; interferon alpha 17; interferon alpha-17; Interferon alpha-88; interferon alpha-I'; Interferon alpha-T; interferon alpha-WA; interferon, alpha 17; leIF I; LEIF2C1
Gene Symbols IFNA17
Host Species Rabbit
Immunogen A synthesized peptide derived from human IFNA17(Accession P01571), corresponding to amino acid residues L40-L90.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3451
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.