Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IFNA4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121885
This item is not returnable.
View return policy
Description
IFNA4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Specifications
IFNA4 | |
Polyclonal | |
Unconjugated | |
IFNA4 | |
Ifa4; IFNA4; IFN-alpha-4; IFN-alpha4a; INFA4; Interferon; interferon alpha 4; interferon alpha family, gene 4; interferon alpha-4; Interferon alpha-4B; interferon alpha-76; Interferon alpha-M1; interferon, alpha 4; MuIFN-alpha 4 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
15967, 298205, 3441 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P05014, P07351 | |
IFNA4 | |
Recombinant protein Interferon Alpha 4. | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction