Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFNA5 Monoclonal Antibody (C2), Invitrogen™

Catalog No. PIMA543698
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543698 20 μL
PIMA543697 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543698 Supplier Invitrogen™ Supplier No. MA543698
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFNA5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene IFNA5
Gene Accession No. P01569
Gene Alias Ifa5; Ifna5; IFN-alpha-5; IFN-alphaG; INA5; INFA5; Interferon; interferon alpha 5; interferon alpha family, gene 5; interferon alpha-5; Interferon alpha-61; interferon alpha-G; interferon, alpha 5; leIF G
Gene Symbols IFNA5
Host Species Mouse
Immunogen Leu22-Glu189 (Accession P01569), with N-terminal His and GST Tag.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3442
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.