Learn More
Invitrogen™ IFNA5 Recombinant Rabbit Monoclonal Antibody (102)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA530505
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human IFNA5/IFNaG.
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Specifications
IFNA5 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P01569 | |
IFNA5 | |
Recombinant Human IFNA5/IFNaG Protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
102 | |
Unconjugated | |
IFNA5 | |
Ifa5; Ifna5; IFN-alpha-5; IFN-alphaG; INA5; INFA5; Interferon; interferon alpha 5; interferon alpha family, gene 5; interferon alpha-5; Interferon alpha-61; interferon alpha-G; interferon, alpha 5; leIF G | |
Rabbit | |
Protein A | |
RUO | |
3442 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.