Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFNA7 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA570101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570101 Supplier Thermo Scientific Supplier No. PA570101

Rabbit Polyclonal Antibody

This target displays homology in the following species: Guinea Pig: 75%; Human: 100%; Mouse: 75%

Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFNA7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene IFNA7
Gene Accession No. P01567
Gene Alias Ifa7; IFNA7; IFNA-J; IFN-alpha 7; IFN-alpha-7; IFN-alphaJ; IFN-alpha-J1; interferon alpha 7; interferon alpha family, gene 7; interferon alpha-7; Interferon alpha-J; interferon alpha-J1; interferon, alpha 7; leIF J
Gene Symbols IFNA7
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human IFNA7
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3444
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.