Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFNAR1 Polyclonal Antibody

Goat Polyclonal Antibody

Supplier:  Invitrogen™ PA124777

Catalog No. PA124777


Only null left
Add to Cart

Description

Description

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Interferons are widely used therapeutic agents because of their anti tumor and antiviral effects and because of their modulatory effects on the immune system (Biron,2001, Kirkwood, 2002). These cytokines produce their effects by binding to the Type 1 Interferon-& Receptor (IFNAR1). Down regulation of this receptor plays a key role in determining the magnitude and duration of cytokine signaling. This down regulation is thought to be influenced by phosphorylation of Serine 535 and 539 in the IFNAR1 (Kumar et al., 2003).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IFNAR1
Polyclonal
Unconjugated
Ifnar1
alpha-type antiviral protein; AVP; beta-type antiviral protein; CD118; CRF2-1; cytokine receptor class-II member 1; Cytokine receptor family 2 member 1; H3K1; Ifar; IFN R1; IFNalpha/beta R1; IFN-alpha/beta receptor 1; IFN-alpha/betaR; IFNalpha/betaR1; IFN-alpha/betaR1; IFN-alpha/betaRalpha; IFNalpha/beta-Ralpha; IFN-alpha-REC; IFNAR; Ifnar1; IFNBR; IFNR1; IFN-R-1; Ifrc; INF-a receptor; Infar; interferon (alpha and beta) receptor 1; interferon (alpha, beta and omega) receptor 1; interferon alpha and beta receptor subunit 1; interferon alpha/beta receptor 1; interferon receptor 1; interferon-alpha/beta receptor 1; interferon-alpha/beta receptor alpha chain; interferon-beta receptor 1; type I interferon receptor 1
Goat
Antigen affinity chromatography
RUO
3454
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Western Blot
0.1 mg/mL
PBS with no preservative
P17181
Ifnar1
Purified, recombinant human interferon-alpha/beta receptor 1 (IFNAR1) extracellular domain expressed in insect Sf cells.
25 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.