Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFNAR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538503
Description
Interferons are widely used therapeutic agents because of their anti tumor and antiviral effects and because of their modulatory effects on the immune system (Biron,2001; Kirkwood, 2002). These cytokines produce their effects by binding to the Type 1 Interferon-& Receptor (IFNAR1). Down regulation of this receptor plays a key role in determining the magnitude and duration of cytokine signaling. This down regulation is thought to be influenced by phosphorylation of Serine 535 and 539 in the IFNAR1 (Kumar et al., 2003).
Specifications
| IFNAR1 | |
| Polyclonal | |
| Unconjugated | |
| IFNAR1 | |
| AVP, IFN-alpha-REC, IFNAR, IFNBR, IFRC, CRF2-1, IFN-R-1, IFN-alpha/beta receptor 1, alpha-type antiviral protein, beta-type antiviral protein, cytokine receptor class-II member 1, cytokine receptor family 2 member 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, type I interferon receptor 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3454 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P17181 | |
| IFNAR1 | |
| A synthetic peptide derived from the internal region of human Interferon-alpha/beta Receptor alpha chain | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction