Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFNAR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102479
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102479 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102479 Supplier Invitrogen™ Supplier No. PA5102479
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IFNAR2.

The type I interferons, IFN-alpha and IFN-beta, are a group of structurally and functionally related proteins that are induced by either viruses or double-stranded RNA and are defined by their ability to confer an antiviral state in cells. IFN-alpha and IFN-beta appear to compete with one another for binding to a common cell surface receptor, while immune IFN (IFN-gamma) binds to a distinct receptor. This distinct receptor, IFN-alphaR, is only weakly responsive to type I interferons, in contrast to IFN-alpha/betaR, which binds to and responds effectively to IFN-beta and to several of the IFN-alpha subtypes. IFN-alpha/betaR is expressed as two alternatively spliced transcripts, designated IFN-alpha/betaRalpha (IFN-alpha/betaR1) and IFN-alpha/betaRbeta (IFN-alpha/betaR2), both of which are involved in signal transduction and ligand binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFNAR2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IFNAR2
Gene Accession No. P48551
Gene Alias AI747302; human interferon alpha/beta receptor; IFN alpha/betaRbeta; IFN R2; IFNABR; IFNalpha/beta R2; IFN-alpha/beta receptor 2; IFNalpha/betaR2; IFN-alpha/betaR2; IFN-alpha/betaRbeta; IFNalpha/beta-Rbeta; IFN-alpha-REC; IFNAR2; Ifnar-2; IFNARB; IFN-R; IFNR2; IFN-R-2; IMD45; interferon (alpha and beta) receptor 2; interferon (alpha, beta and omega) receptor 2; interferon alpha and beta receptor subunit 2; interferon alpha binding protein; Interferon alpha/beta receptor 2; interferon receptor 2c; interferon-alpha/beta receptor 2; interferon-alpha/beta receptor beta chain; type I interferon alpha/beta receptor 2a; type I interferon receptor 2
Gene Symbols IFNAR2
Host Species Rabbit
Immunogen A synthesized peptide derived from human IFNAR2(Accession P48551), corresponding to amino acid residues L453-E503.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3455
Target Species Human, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.