Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFNAR2 Recombinant Rabbit Monoclonal Antibody (117)
GREENER_CHOICE

Catalog No. PIMA530508
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530508 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530508 Supplier Invitrogen™ Supplier No. MA530508
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human IFNAR2.

The type I interferons, IFN-alpha and IFN-beta, are a group of structurally and functionally related proteins that are induced by either viruses or double-stranded RNA and are defined by their ability to confer an antiviral state in cells. IFN-alpha and IFN-beta appear to compete with one another for binding to a common cell surface receptor, while immune IFN (IFN-gamma) binds to a distinct receptor. This distinct receptor, IFN-alphaR, is only weakly responsive to type I interferons, in contrast to IFN-alpha/betaR, which binds to and responds effectively to IFN-beta and to several of the IFN-alpha subtypes. IFN-alpha/betaR is expressed as two alternatively spliced transcripts, designated IFN-alpha/betaRalpha (IFN-alpha/betaR1) and IFN-alpha/betaRbeta (IFN-alpha/betaR2), both of which are involved in signal transduction and ligand binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFNAR2
Applications ELISA
Classification Recombinant Monoclonal
Clone 117
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene IFNAR2
Gene Accession No. P48551
Gene Alias AI747302; human interferon alpha/beta receptor; IFN alpha/betaRbeta; IFN R2; IFNABR; IFNalpha/beta R2; IFN-alpha/beta receptor 2; IFNalpha/betaR2; IFN-alpha/betaR2; IFN-alpha/betaRbeta; IFNalpha/beta-Rbeta; IFN-alpha-REC; IFNAR2; Ifnar-2; IFNARB; IFN-R; IFNR2; IFN-R-2; IMD45; interferon (alpha and beta) receptor 2; interferon (alpha, beta and omega) receptor 2; interferon alpha and beta receptor subunit 2; interferon alpha binding protein; Interferon alpha/beta receptor 2; interferon receptor 2c; interferon-alpha/beta receptor 2; interferon-alpha/beta receptor beta chain; type I interferon alpha/beta receptor 2a; type I interferon receptor 2
Gene Symbols IFNAR2
Host Species Rabbit
Immunogen Recombinant Human IFNAR2 protein (Met1-Lys243).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3455
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.