Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFNAR2 Recombinant Rabbit Monoclonal Antibody (122), APC
GREENER_CHOICE

Catalog No. PIMA540952
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA540952 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA540952 Supplier Invitrogen™ Supplier No. MA540952
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

May be stored for twelve months without detectable loss of activity. Protect from prolonged exposure to light and do not freeze. Sodium azide is toxic to cells and should be disposed of properly so flush with large volumes of water during disposal.

The type I interferons, IFN-alpha and IFN-beta, are a group of structurally and functionally related proteins that are induced by either viruses or double-stranded RNA and are defined by their ability to confer an antiviral state in cells. IFN-alpha and IFN-beta appear to compete with one another for binding to a common cell surface receptor, while immune IFN (IFN-gamma) binds to a distinct receptor. This distinct receptor, IFN-alphaR, is only weakly responsive to type I interferons, in contrast to IFN-alpha/betaR, which binds to and responds effectively to IFN-beta and to several of the IFN-alpha subtypes. IFN-alpha/betaR is expressed as two alternatively spliced transcripts, designated IFN-alpha/betaRalpha (IFN-alpha/betaR1) and IFN-alpha/betaRbeta (IFN-alpha/betaR2), both of which are involved in signal transduction and ligand binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFNAR2
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 122
Concentration 0.1 mg/mL
Conjugate APC
Formulation PBS with 0.5% BSA and 0.03% ProClin 300
Gene IFNAR2
Gene Accession No. P48551
Gene Alias AI747302; human interferon alpha/beta receptor; IFN alpha/betaRbeta; IFN R2; IFNABR; IFNalpha/beta R2; IFN-alpha/beta receptor 2; IFNalpha/betaR2; IFN-alpha/betaR2; IFN-alpha/betaRbeta; IFNalpha/beta-Rbeta; IFN-alpha-REC; IFNAR2; Ifnar-2; IFNARB; IFN-R; IFNR2; IFN-R-2; IMD45; interferon (alpha and beta) receptor 2; interferon (alpha, beta and omega) receptor 2; interferon alpha and beta receptor subunit 2; interferon alpha binding protein; Interferon alpha/beta receptor 2; interferon receptor 2c; interferon-alpha/beta receptor 2; interferon-alpha/beta receptor beta chain; type I interferon alpha/beta receptor 2a; type I interferon receptor 2
Gene Symbols IFNAR2
Host Species Rabbit
Immunogen Recombinant Human IFNAR2 Protein.
Purification Method Protein A
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3455
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.