Learn More
IFNG Mouse anti-Human, Clone: SPM153, Abnova™
Mouse monoclonal antibody raised against full length recombinant human IFNG.
Supplier: Abnova Corporation MAB13091
Description
Interferon-gamma (IFNG), or type II interferon, is a cytokine critical for innate and adaptive immunity against viral and intracellular bacterial infections and for tumor control. Aberrant IFNG expression is associated with a number of autoinflammatory and autoimmune diseases. The importance of IFNG in the immune system stems in part from its ability to inhibit viral replication directly, but most importantly derives from its immunostimulatory and immunomodulatory effects. IFNG is produced predominantly by natural killer (NK) and natural killer T (NKT) cells as part of the innate immune response, and by CD4 (MIM 186940) and CD8 (see MIM 186910) cytotoxic T lymphocyte (CTL) effector T cells once antigen-specific immunity develops (Schoenborn and Wilson, 2007 [PubMed 17981204]).[supplied by OMIM]
Specifications
| IFNG | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| IFG/IFI | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 3458 | |
| Store at 4°C. | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence | |
| SPM153 | |
| Mouse monoclonal antibody raised against full length recombinant human IFNG. | |
| In 10mM PBS (0.05% BSA, 0.05% sodium azide). | |
| IFNG | |
| Recombinant protein corresponding to full length human IFNG. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.