Learn More
Invitrogen™ IFNGR1 (CD119) Recombinant Rat Monoclonal Antibody (GR20)

Description
Specificity: This antibody recognises IFN-gammaRalpha, which is a class II cytokine receptor family member that serves as a IFN-gamma-binding chain associated with the IFN-gamma Beta.
Specifications
Specifications
| Antigen | IFNGR1 (CD119) |
| Applications | Flow Cytometry, Inhibition Assays, Neutralization |
| Classification | Recombinant Monoclonal |
| Clone | GR20 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | IFNGR1 |
| Gene Accession No. | P15261 |
| Gene Alias | antiviral protein, type 2; AVP, type 2; CD119; CD119 antigen; CDw119; Ifgr; IFN-gamma receptor 1; IFN-gammaR; IFN-gamma-R1; IFN-gamma-R-alpha; Ifngr; IFNGR1; IMD27A; IMD27B; immune interferon receptor 1; INF-g receptor; interferon gamma receptor 1; Interferon gamma receptor alpha-chain; interferon-gamma receptor 1; interferon-gamma receptor alpha chain; Nktar |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.