Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IFNGR2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587009
Description
Purity is > 95% by SDS-PAGE.
This gene (IFNGR2) encodes the non-ligand-binding beta chain of the gamma interferon receptor. Human interferon-gamma receptor is a heterodimer of IFNGR1 and IFNGR2. Defects in IFNGR2 are a cause of mendelian susceptibility to mycobacterial disease (MSMD), also known as familial disseminated atypical mycobacterial infection. MSMD is a genetically heterogeneous disease with autosomal recessive, autosomal dominant or X-linked inheritance.
Specifications
IFNGR2 | |
Polyclonal | |
Unconjugated | |
IFNGR2 | |
AF-1; Ifgr2; Ifgt; IFN-gamma receptor 2; IFN-gamma-R2; IFN-gamma-R-beta; IFNGR2; IFNGT1; IMD28; Interferon gamma receptor 2; interferon gamma receptor 2 (interferon gamma transducer 1); interferon gamma receptor accessory factor 1; interferon gamma receptor accessory factor-1; interferon gamma receptor beta chain; Interferon gamma receptor beta-chain; Interferon gamma transducer 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15980, 3460, 360697 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P38484 | |
IFNGR2 | |
Synthetic peptide of human IFNGR2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction