Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFNK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545332
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545332 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545332 Supplier Invitrogen™ Supplier No. PA545332
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MKPSEARVPQ LSSLELRRYF HRIDNFLKEK KYSDCAWEIV RVEIRRCLYY Sequence homology: Cow: 85%; Dog: 93%; Goat: 77%; Guinea Pig: 85%; Horse: 93%; Human: 100%; Pig: 92%; Rat: 77%; Sheep: 85%.

This gene encodes a member of the type I interferon family. Type I interferons are a group of related glycoproteins that play an important role in host defenses against viral infections. This protein is expressed in keratinocytes and the gene is found on chromosome 9, adjacent to the type I interferon cluster.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFNK
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene IFNK
Gene Accession No. Q9P0W0
Gene Alias Ifnk; IFN-kappa; IFNT1; INFE1; interferon; Interferon kappa; interferon, kappa; interferon-like protein; RP11-27J8.1; type I interferon kappa; UNQ6124/PRO20084
Gene Symbols IFNK
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human IFNK (aa 147-196).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56832
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.