Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IFNK Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103430
Description
Antibody detects endogenous levels of total IFNK.
This gene encodes a member of the type I interferon family. Type I interferons are a group of related glycoproteins that play an important role in host defenses against viral infections. This protein is expressed in keratinocytes and the gene is found on chromosome 9, adjacent to the type I interferon cluster.
Specifications
| IFNK | |
| Polyclonal | |
| Unconjugated | |
| IFNK | |
| Ifnk; IFN-kappa; IFNT1; INFE1; interferon; Interferon kappa; interferon, kappa; interferon-like protein; RP11-27J8.1; type I interferon kappa; UNQ6124/PRO20084 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 56832 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9P0W0 | |
| IFNK | |
| A synthesized peptide derived from human IFNK(Accession Q9P0W0), corresponding to amino acid residues Q113-R163. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction