Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IFNL4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144239
Description
IFNL4 Polyclonal Antibody for ICC/IF, ELISA
Cytokine that may trigger an antiviral response activating the JAK-STAT pathway and up-regulating specifically some interferon-stimulated genes.
Specifications
IFNL4 | |
Polyclonal | |
Unconjugated | |
IFNL4 | |
IFNAN; IFNL4; IFNL4_HUMAN; IFN-lambda-4; Interferon; Interferon lambda-4 | |
Rabbit | |
Protein G | |
RUO | |
101180976 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunocytochemistry | |
3.5 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
K9M1U5 | |
IFNL4 | |
Recombinant Human Interferon lambda-4 protein (34-76AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction