Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFRD1 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299593APC
Description
IFRD1 Polyclonal antibody specifically detects IFRD1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
IFRD1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
12-O-tetradecanoylphorbol-13-acetate-induced sequence 7, interferon-related developmental regulator 1, Nerve growth factor-inducible protein PC4, PC4, pheochromocytoma cell-4, TIS7, TPA induced sequence 7 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human IFRD1. | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication | |
3475 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction