Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFRD1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA528060

Catalog No. PIPA528060


Only null left
Add to Cart

Description

Description

Recommended positive controls: Molt-4, Raji. Predicted reactivity: Mouse (94%), Rat (94%), Pig (94%), Chicken (81%), Rhesus Monkey (99%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Tis7, also known as IFRD1, is a 442 amino acid containing membrane associated, non-nuclear intracellular protein belonging to the IFRD family. A regulatory protein expressed in a variety of tissues, Tis7 is known to play an important role in the regulation of gene activity in the proliferative and/or differentiative pathways induced by NGF. Reports suggest that Tis7 may be an autocrine factor that attenuates or amplifies the initial ligand induced signal and might also augment the adaptive response by stimulating the production of differentiated enterocytes. Tis7 acts as a negative regulator of transcriptional activity and represses expression of genes involved in myogenesis, muscle maintenance, and regeneration in a histone deacetylase dependent manner.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IFRD1
Polyclonal
Unconjugated
IFRD1
12-O-tetradecanoylphorbol-13-acetate-induced sequence 7; hypothetical protein LOC541283; Ifnl; Ifrd1; interferon related developmental regulator 1; interferon-related developmental regulator 1; IRPR; nerve growth factor-inducible protein PC4; Pc4; pheochromocytoma cell-4; Tis7; TIS7 protein; TPA induced sequence 7; TPA-induced sequence 7
Rabbit
Antigen affinity chromatography
RUO
3475
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.62 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O00458
IFRD1
Recombinant protein encompassing a sequence within the center region of human IFRD1. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.