Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFRD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554985
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554985 Supplier Invitrogen™ Supplier No. PA554985
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AKVDKRKQRS VFRDVLRAVE ERDFPTETIK FGPERMYIDC WVKKHTYDTF KEVLGSGMQY HLQSNEFLRN VFELGPPVML DAATLK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 95%.

Tis7, also known as IFRD1, is a 442 amino acid containing membrane associated, non-nuclear intracellular protein belonging to the IFRD family. A regulatory protein expressed in a variety of tissues, Tis7 is known to play an important role in the regulation of gene activity in the proliferative and/or differentiative pathways induced by NGF. Reports suggest that Tis7 may be an autocrine factor that attenuates or amplifies the initial ligand induced signal and might also augment the adaptive response by stimulating the production of differentiated enterocytes. Tis7 acts as a negative regulator of transcriptional activity and represses expression of genes involved in myogenesis, muscle maintenance, and regeneration in a histone deacetylase dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFRD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IFRD1
Gene Accession No. O00458
Gene Alias 12-O-tetradecanoylphorbol-13-acetate-induced sequence 7; hypothetical protein LOC541283; Ifnl; Ifrd1; interferon related developmental regulator 1; interferon-related developmental regulator 1; IRPR; nerve growth factor-inducible protein PC4; Pc4; pheochromocytoma cell-4; Tis7; TIS7 protein; TPA induced sequence 7; TPA-induced sequence 7
Gene Symbols IFRD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IFRD1. Recombinant protein control fragment (Product #RP-92274).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3475
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.