Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFT122 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297508MFV450
Description
IFT122 Polyclonal antibody specifically detects IFT122 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
IFT122 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
intraflagellar transport 122 homolog (Chlamydomonas), intraflagellar transport protein 122 homolog, SPGWD repeat-containing protein 140, WD repeat domain 10, WD repeat-containing protein 10, WDR10WDR10p, WDR140CED | |
Produced in rabbits immunized with E. coli-derived Human IFT122 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
55764 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction