Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFT122 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100568
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100568 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100568 Supplier Invitrogen™ Supplier No. PA5100568
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IFT122.

This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This cytoplasmic protein contains seven WD repeats and an AF-2 domain which function by recruiting coregulatory molecules and in transcriptional activation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFT122
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IFT122
Gene Accession No. Q6NWV3, Q9HBG6
Gene Alias C86139; CED; CED1; hypothetical protein zgc:77217; Ift122; intraflagellar transport 122; intraflagellar transport 122 homolog; intraflagellar transport 122 homolog (Chlamydomonas); intraflagellar transport protein 122 homolog; sopb; SPG; WD repeat domain 10; WD repeat-containing protein 10; WD repeat-containing protein 140; WDR10; WDR10p; WDR140; zgc:77217
Gene Symbols IFT122
Host Species Rabbit
Immunogen A synthesized peptide derived from human IFT122(Accession Q9HBG6), corresponding to amino acid residues I87-C137.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55764, 81896
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.