Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFT140 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559910
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559910 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559910 Supplier Invitrogen™ Supplier No. PA559910
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LWRLDQRGRV QGTPLLKHEY GKHLTHCIFR LPPPGEDLVQ LAKAAVSGDE KALDMFNWKK SSSGSLLKMG SHEGLLFFVS LMDGTVHYVD EKGKTTQVVS ADSTIQMLFY Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

IFT140 contains 9 TPR repeats and 5 WD repeats. The function of the IFT140 protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFT140
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IFT140
Gene Accession No. Q96RY7
Gene Alias AI661311; c305C8.4; c380F5.1; gs114; Ift140; intraflagellar transport 140; intraflagellar transport 140 homolog; intraflagellar transport protein 140 homolog; KIAA0590; LOW QUALITY PROTEIN: intraflagellar transport protein 140 homolog; mKIAA0590; MZSDS; SRTD9; Tce5; T-complex expressed gene 5; WD and tetratricopeptide repeats 2; WD and tetratricopeptide repeats protein 2; WDTC2
Gene Symbols IFT140
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IFT140. Recombinant protein control fragment (Product #RP-98798).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9742
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.