Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFT172 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560861
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560861 Supplier Invitrogen™ Supplier No. PA560861
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSPGTNCAEA YHSWADLRDV LFNLCENLVK SSEANSPAHE EFKTMLLIAH YYATRSAAQS VKQLETVAAR LSVSLLRHTQ LLPVDKAFYE AGIAAKAVGW DNMAFIFLNR Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.

IFT172 is required for the maintenance and formation of cilia. IT plays an indirect role in hedgehog (Hh) signaling, cilia being required for all activity of the hedgehog pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFT172
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IFT172
Gene Accession No. Q9UG01
Gene Alias 4930553F24Rik; avc1; BBS20; ift172; intraflagellar transport 172; intraflagellar transport 172 homolog; intraflagellar transport 172 protein; intraflagellar transport protein 172 homolog; Kiaa1179; LIM-interacting protein; moe; NPHP17; osm-1; Protein wimple; RP71; selective LIM binding factor homolog; selective LIM binding factor, rat homolog; selective LIM-binding factor; selective LIM-binding factor Wimple; si:dkey-221h15.5; Slb; SRTD10; Wim; wimple homolog
Gene Symbols IFT172
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IFT172. Recombinant protein control fragment (Product #RP-91636).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26160
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.