Learn More
Invitrogen™ IFT20 Polyclonal Antibody

Description
This antibody is predicted to react with bovine based on sequence homology.
Specifications
Specifications
| Antigen | IFT20 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | IFT20 |
| Gene Accession No. | Q61025, Q8IY31 |
| Gene Alias | 0610009H04Rik; AU015496; hIFT20; IFT20; intraflagellar transport 20; intraflagellar transport 20 homolog; intraflagellar transport protein 20 homolog; intraflagellar transport protein IFT20; mIFT20; RGD1309400; uterine protein |
| Gene Symbols | IFT20 |
| Host Species | Rabbit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.