Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IFT20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554394
Description
Immunogen sequence: MTHLLLTATV TPSEQNSSRE PGWETAMAKD ILGEAGLHFD ELNKLRVLDP EVTQQTIELK EECKDFVDK Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 87%.
IFT20 is part of intraflagellar transport (IFT) particles involved in ciliary process assembly. IFT20 may play a role in the trafficking of ciliary membrane proteins from the Golgi complex to the cilium.
Specifications
| IFT20 | |
| Polyclonal | |
| Unconjugated | |
| IFT20 | |
| 0610009H04Rik; AU015496; hIFT20; IFT20; intraflagellar transport 20; intraflagellar transport 20 homolog; intraflagellar transport protein 20 homolog; intraflagellar transport protein IFT20; mIFT20; RGD1309400; uterine protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90410 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IY31 | |
| IFT20 | |
| Recombinant protein corresponding to Human IFT20. Recombinant protein control fragment (Product #RP-93222). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction