Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFT20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112312
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112312 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112312 Supplier Invitrogen™ Supplier No. PA5112312
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IFT20 Polyclonal Antibody for Western Blot, IHC (P), IP

IFT20 is part of intraflagellar transport (IFT) particles involved in ciliary process assembly. IFT20 may play a role in the trafficking of ciliary membrane proteins from the Golgi complex to the cilium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFT20
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene IFT20
Gene Accession No. Q8IY31
Gene Alias 0610009H04Rik; AU015496; hIFT20; IFT20; intraflagellar transport 20; intraflagellar transport 20 homolog; intraflagellar transport protein 20 homolog; intraflagellar transport protein IFT20; mIFT20; RGD1309400; uterine protein
Gene Symbols IFT20
Host Species Rabbit
Immunogen E. coli-derived Human IFT20/intraflagellar transport 20 fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90410
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.