Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFT25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100040
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100040 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100040 Supplier Invitrogen™ Supplier No. PA5100040
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IFT25.

HSPB11 (Heat shock protein beta-11), also known as IFT25, was originally identified as a small heat shock protein. Recently it has been identified as a component of IFT complex B. It has been demonstrated that IFT25 is not required for ciliary assembly but is required for proper Hedgehog signaling, which in mammals occurs within cilia. Cilia lacking IFT25 have defects in the signal-dependent transport of multiple Hedgehog components and fail to activate the pathway upon stimulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFT25
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Hspb11
Gene Accession No. Q9D6H2, Q9Y547
Gene Alias 2900042B11Rik; C1orf41; Heat shock protein; Heat shock protein beta-11; heat shock protein family B (small) member 11; heat shock protein family B (small), member 11; HSP; Hspb11; Hspb11 heat shock protein family B (small), member 11; HSPC034; HSPCO34; IFT25; intraflagellar transport 25 homolog; intraflagellar transport protein 25 homolog; Placental protein 25; placental protein 25 homolog; PP25
Gene Symbols Hspb11
Host Species Rabbit
Immunogen A synthesized peptide derived from human HSPB11(Accession Q9Y547), corresponding to amino acid residues C7-V57.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51668, 685284, 72938
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.