Learn More
IFT25 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 157321AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0439
Only null left
Add to Cart
Description
Description
HSPB11 (Heat shock protein beta-11), also known as IFT25, was originally identified as a small heat shock protein. Recently it has been identified as a component of IFT complex B. It has been demonstrated that IFT25 is not required for ciliary assembly but is required for proper Hedgehog signaling, which in mammals occurs within cilia. Cilia lacking IFT25 have defects in the signal-dependent transport of multiple Hedgehog components and fail to activate the pathway upon stimulation.
Specifications
Specifications
IFT25
Polyclonal
Unconjugated
HSPB11
C1orf41, Heat shock protein beta 11, HSPB11, HSPCO34, IFT25, ITF25, Placental protein 25, PP25
Rabbit
Antigen Affinity Chromatography
RUO
51668, 685284, 72938
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9D6H2, Q9Y547
HSPB11
IFT25 Fusion Protein Ag8490
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.