Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IFT46 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566089
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566089 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566089 Supplier Invitrogen™ Supplier No. PA566089
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDEPSTKQSDP TVLSLWLTEN SKQHNITQHM KVKSLEDAEK NPKAIDTWIE SISELHRSKP PATVHYTRPM PDIDTLMQEW SPEF Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

Forms part of a complex involved in intraflagellar transport (IFT), the bi-directional movement of particles required for the assembly, maintenance and functioning of primary cilia. May play a role in chondrocyte maturation and skeletogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFT46
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IFT46
Gene Accession No. Q9NQC8
Gene Alias 1500035H01Rik; AA408110; AI663846; c11orf2; C11orf60; CFAP32; cilia and flagella associated protein 32; Ift46; intraflagellar transport 46; intraflagellar transport 46 homolog; intraflagellar transport protein 46 homolog; intraflagellar transport protein IFT46; R75047; UPF0360
Gene Symbols IFT46
Host Species Rabbit
Immunogen Recombinant Human IFT46. Recombinant protein control fragment (Product #RP-106785).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56912
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.